Product Details
- SNP ID
-
rs144879123
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:35700020 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCAGGTAGCCATTGGCACACTCC[C/T]GGCCATAGTGCAGGGCTCGAAGCCG
- Phenotype
-
MIM: 186745
MIM: 190990
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TLN1
PubMed Links
Gene Details
- Gene
- TLN1
- Gene Name
- talin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006289.3 |
7076 |
Missense Mutation |
CAG,CGG |
Q2241R |
NP_006280.3 |
- Gene
- TPM2
- Gene Name
- tropomyosin 2 (beta)
There are no transcripts associated with this gene.
View Full Product Details