Product Details

SNP ID
rs145131863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:98060868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCTTGGAGAGGCCATACACCTC[A/G]AAGCATTCCTGGGGGAAGACGTACG
Phenotype
MIM: 605202
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NANS PubMed Links

Gene Details

Gene
NANS
Gene Name
N-acetylneuraminate synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018946.3 351 Silent Mutation TCA,TCG S73S NP_061819.2
XM_011518787.2 351 Intron XP_011517089.1
XM_011518788.2 351 Intron XP_011517090.1
XM_017014811.1 351 Intron XP_016870300.1

View Full Product Details