Product Details
- SNP ID
-
rs145825708
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:21333136 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGACTCTGGAAGATCAAAAACTTTA[C/T]GCCACTCATCTTTTTCTGGGTCATA
- Phenotype
-
MIM: 611201
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KLHL9
PubMed Links
Gene Details
- Gene
- KLHL9
- Gene Name
- kelch like family member 9
There are no transcripts associated with this gene.
View Full Product Details