Product Details

SNP ID
rs146291236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:83663882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAAAAGAACTGATACCTGAGGA[C/T]TTACTCCAGCAAGAGCCTGCAGCAT
Phenotype
MIM: 605046
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBQLN1 PubMed Links

Gene Details

Gene
UBQLN1
Gene Name
ubiquilin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013438.4 2100 Missense Mutation NP_038466.2
NM_053067.2 2100 Missense Mutation NP_444295.1
XM_005251948.2 2100 Intron XP_005252005.1

View Full Product Details