Product Details

SNP ID
rs147089074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:132375990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCATGCATGGCGCCTGGCCTTC[A/G]CTTTCTTTTTCTATGTCCTCTCCTT
Phenotype
MIM: 600777
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTF1 PubMed Links

Gene Details

Gene
TTF1
Gene Name
transcription termination factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205296.1 3219 Silent Mutation NP_001192225.1
NM_007344.3 3219 Silent Mutation NP_031370.2
XM_006717273.3 3219 Silent Mutation XP_006717336.2
XM_017015105.1 3219 Silent Mutation XP_016870594.1

View Full Product Details