Product Details

SNP ID
rs147332575
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:108935688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTGTATACTACAGCACTCTAAC[C/T]GGTAAGCAAATTGGGAATTTTAAAT
Phenotype
MIM: 604785 MIM: 603722
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CTNNAL1 PubMed Links

Gene Details

Gene
CTNNAL1
Gene Name
catenin alpha like 1
There are no transcripts associated with this gene.

Gene
FAM206A
Gene Name
family with sequence similarity 206 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017832.3 220 Nonsense Mutation CGA,TGA R44* NP_060302.1
XM_011518816.2 220 Nonsense Mutation CGA,TGA R44* XP_011517118.1
Gene
IKBKAP
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
There are no transcripts associated with this gene.

View Full Product Details