Product Details

SNP ID
rs147462165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136484775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTTGCCTTTCAGGAGTCAGATT[C/T]CTGACCTGTTTGAACAGAGTTAGGG
Phenotype
MIM: 190198 MIM: 612854
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf163 PubMed Links

Gene Details

Gene
C9orf163
Gene Name
chromosome 9 open reading frame 163
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152571.2 1281 Silent Mutation TTC,TTT F109F NP_689784.1
Gene
NOTCH1
Gene Name
notch 1
There are no transcripts associated with this gene.

Gene
SEC16A
Gene Name
SEC16 homolog A, endoplasmic reticulum export factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276418.1 1281 Intron NP_001263347.1
NM_014866.1 1281 Intron NP_055681.1
XM_005266134.2 1281 Intron XP_005266191.1
XM_011519246.2 1281 Intron XP_011517548.1
XM_011519247.1 1281 Intron XP_011517549.1
XM_011519248.2 1281 Intron XP_011517550.1
XM_011519249.1 1281 Intron XP_011517551.1
XM_011519250.2 1281 Intron XP_011517552.1
XM_011519251.1 1281 Intron XP_011517553.1
XM_011519252.2 1281 Intron XP_011517554.1
XM_011519253.1 1281 Intron XP_011517555.1
XM_011519254.1 1281 Intron XP_011517556.1
XM_011519255.2 1281 Intron XP_011517557.1
XM_011519256.1 1281 Intron XP_011517558.1
XM_011519257.1 1281 Intron XP_011517559.1
XM_011519258.1 1281 Intron XP_011517560.1
XM_011519259.1 1281 Intron XP_011517561.1
XM_011519260.2 1281 Intron XP_011517562.1
XM_011519261.1 1281 Intron XP_011517563.1
XM_011519262.1 1281 Intron XP_011517564.1
XM_011519263.2 1281 Intron XP_011517565.1
XM_011519264.2 1281 Intron XP_011517566.1
XM_017015363.1 1281 Intron XP_016870852.1
XM_017015364.1 1281 Intron XP_016870853.1

View Full Product Details