Product Details

SNP ID
rs147773784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120858253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCAGGGGCATGTATGCCTTGGC[A/T]GCCAGCTTCCTCTTTCGGCTGCCCA
Phenotype
MIM: 609740
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PHF19 PubMed Links

Gene Details

Gene
PHF19
Gene Name
PHD finger protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009936.2 1558 Intron NP_001009936.1
NM_001286840.1 1558 Silent Mutation GCA,GCT A497A NP_001273769.1
NM_001286842.1 1558 Silent Mutation GCA,GCT A269A NP_001273771.1
NM_001286843.1 1558 Intron NP_001273772.1
NM_015651.2 1558 Silent Mutation GCA,GCT A478A NP_056466.1
XM_005251906.2 1558 Silent Mutation GCA,GCT A478A XP_005251963.1
XM_011518509.2 1558 Silent Mutation GCA,GCT A478A XP_011516811.1
XM_011518511.2 1558 UTR 3 XP_011516813.1
XM_011518515.2 1558 Intron XP_011516817.1
XM_011518516.2 1558 Intron XP_011516818.1
XM_017014612.1 1558 Silent Mutation GCA,GCT A478A XP_016870101.1
XM_017014613.1 1558 Intron XP_016870102.1
Gene
PSMD5-AS1
Gene Name
PSMD5 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details