Product Details
- SNP ID
-
rs148110858
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:104695246 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAAAGCCTTCTCTACCTGTTCAGC[A/G]CACTCGATTGTGGTCATCTTATTCT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR13D1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10761073] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR13D1
- Gene Name
- olfactory receptor family 13 subfamily D member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004484.1 |
825 |
Silent Mutation |
GCA,GCG |
A275A |
NP_001004484.1 |
View Full Product Details