Product Details

SNP ID
rs148328690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133344272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTCGTTCACGGAGGGGAAGTCA[C/T]TGAGGATCAGGAACTGCTTGAGGTC
Phenotype
MIM: 185641 MIM: 185640 MIM: 185620 MIM: 185642
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED22 PubMed Links

Gene Details

Gene
MED22
Gene Name
mediator complex subunit 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133640.4 539 Missense Mutation AAT,AGT N89S NP_598395.1
NM_181491.2 539 Missense Mutation AAT,AGT N89S NP_852468.1
Gene
RPL7A
Gene Name
ribosomal protein L7a
There are no transcripts associated with this gene.

Gene
SNORD24
Gene Name
small nucleolar RNA, C/D box 24
There are no transcripts associated with this gene.

Gene
SNORD36A
Gene Name
small nucleolar RNA, C/D box 36A
There are no transcripts associated with this gene.

Gene
SNORD36B
Gene Name
small nucleolar RNA, C/D box 36B
There are no transcripts associated with this gene.

Gene
SNORD36C
Gene Name
small nucleolar RNA, C/D box 36C
There are no transcripts associated with this gene.

Gene
SURF1
Gene Name
surfeit 1
There are no transcripts associated with this gene.

Gene
SURF6
Gene Name
surfeit 6
There are no transcripts associated with this gene.

View Full Product Details