Product Details

SNP ID
rs148803149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120858222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCATCCAGCTCAGCTGCCCACC[A/G]CTTTGCCCGCAGGGGCATGTATGCC
Phenotype
MIM: 609740
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHF19 PubMed Links

Gene Details

Gene
PHF19
Gene Name
PHD finger protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009936.2 1589 Intron NP_001009936.1
NM_001286840.1 1589 Missense Mutation CGG,TGG R508W NP_001273769.1
NM_001286842.1 1589 Missense Mutation CGG,TGG R280W NP_001273771.1
NM_001286843.1 1589 Intron NP_001273772.1
NM_015651.2 1589 Missense Mutation CGG,TGG R489W NP_056466.1
XM_005251906.2 1589 Missense Mutation CGG,TGG R489W XP_005251963.1
XM_011518509.2 1589 Missense Mutation CGG,TGG R489W XP_011516811.1
XM_011518511.2 1589 UTR 3 XP_011516813.1
XM_011518515.2 1589 Intron XP_011516817.1
XM_011518516.2 1589 Intron XP_011516818.1
XM_017014612.1 1589 Missense Mutation CGG,TGG R489W XP_016870101.1
XM_017014613.1 1589 Intron XP_016870102.1
Gene
PSMD5-AS1
Gene Name
PSMD5 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details