Product Details

SNP ID
rs149157458
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104598735 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTCCCCTCCGAAGAGCTAATTT[C/T]GAAGATGCTCAAAATGATTAACGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR13C2 PubMed Links

Gene Details

Gene
OR13C2
Gene Name
olfactory receptor family 13 subfamily C member 2
There are no transcripts associated with this gene.

Gene
OR13C5
Gene Name
olfactory receptor family 13 subfamily C member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004482.1 679 Missense Mutation AAA,GAA K227E NP_001004482.1

View Full Product Details