Product Details

SNP ID
rs149199268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:20345972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTGAAAGAGCATCCACGGGACC[A/T]AGTACTTATAATGTCTGATGGTGTC
Phenotype
MIM: 159558
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MLLT3 PubMed Links

Gene Details

Gene
MLLT3
Gene Name
MLLT3, super elongation complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286691.1 2232 UTR 3 NP_001273620.1
NM_004529.3 2232 UTR 3 NP_004520.2
XM_017014726.1 2232 UTR 3 XP_016870215.1
XM_017014727.1 2232 UTR 3 XP_016870216.1

View Full Product Details