Product Details

SNP ID
rs150652735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33255165 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGGTAAATTCCGCTCCAGAGAC[A/G]GCAGAGCTGGTGGCGCCATTCTTCA
Phenotype
MIM: 601497 MIM: 610900 MIM: 613929
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BAG1 PubMed Links

Gene Details

Gene
BAG1
Gene Name
BCL2 associated athanogene 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172415.1 1179 UTR 3 NP_001165886.1
NM_004323.5 1179 UTR 3 NP_004314.5
Gene
CHMP5
Gene Name
charged multivesicular body protein 5
There are no transcripts associated with this gene.

Gene
SPINK4
Gene Name
serine peptidase inhibitor, Kazal type 4
There are no transcripts associated with this gene.

View Full Product Details