Product Details

SNP ID
rs150805048
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37424910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAAGGAGCTAGAGCGAGGTGTGG[C/T]GGGGGCCCACGGCCTGCTCTGCCTC
Phenotype
MIM: 604296
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GRHPR PubMed Links

Gene Details

Gene
GRHPR
Gene Name
glyoxylate reductase/hydroxypyruvate reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012203.1 204 Missense Mutation GCG,GTG A50V NP_036335.1
XM_005251631.1 204 Intron XP_005251688.1
XM_011518073.2 204 UTR 5 XP_011516375.1
XM_017015320.1 204 Missense Mutation GCG,GTG A50V XP_016870809.1
XM_017015321.1 204 Missense Mutation GCG,GTG A50V XP_016870810.1
XM_017015322.1 204 Intron XP_016870811.1
XM_017015323.1 204 UTR 5 XP_016870812.1

View Full Product Details