Product Details
- SNP ID
-
rs151217472
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:21332986 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATTAGATCACCCATGACGTACTGC[A/C]AAGGTGTTACACCTTAGACCTAAGA
- Phenotype
-
MIM: 611201
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KLHL9
PubMed Links
Gene Details
- Gene
- KLHL9
- Gene Name
- kelch like family member 9
There are no transcripts associated with this gene.
View Full Product Details