Product Details

SNP ID
rs139185293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:40635382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCTGGAATGGTTAATACAGGCA[C/T]AGTTATCTGCAAGGAGCACAGCTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXorf38 PubMed Links

Gene Details

Gene
CXorf38
Gene Name
chromosome X open reading frame 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144970.2 Intron NP_659407.1
XM_005272589.2 Intron XP_005272646.2
XM_006724527.3 Intron XP_006724590.2
XM_006724528.2 Intron XP_006724591.1
XM_017029302.1 Intron XP_016884791.1
XM_017029303.1 Intron XP_016884792.1
XM_017029304.1 Intron XP_016884793.1
XM_017029305.1 Intron XP_016884794.1

View Full Product Details