Product Details

SNP ID
rs139688762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:140784003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTATCCATGTCTTCCAGAAAAT[C/G]CTTGTCTTCCCTTAAATCTATAGCT
Phenotype
MIM: 302650
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDR1 PubMed Links

Gene Details

Gene
CDR1
Gene Name
cerebellar degeneration related protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004065.2 556 Missense Mutation CAT,GAT H122D NP_004056.2

View Full Product Details