Product Details

SNP ID
rs140427727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:31071811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTAGCGGAAGAAGTTCTCCAGG[A/G]CCACGTCGTCCCGGTTGAAGTAGAA
Phenotype
MIM: 300308
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FTHL17 PubMed Links

Gene Details

Gene
FTHL17
Gene Name
ferritin heavy chain like 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031894.2 Intron NP_114100.1

View Full Product Details