Product Details

SNP ID
rs141220978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:40628923 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCTGCCTGTCCACTGGTCACTC[A/G]AGTCACCATTCTCCAGCTCCCCACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXorf38 PubMed Links

Gene Details

Gene
CXorf38
Gene Name
chromosome X open reading frame 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144970.2 3410 UTR 3 NP_659407.1
XM_005272589.2 3410 Intron XP_005272646.2
XM_006724527.3 3410 UTR 3 XP_006724590.2
XM_006724528.2 3410 UTR 3 XP_006724591.1
XM_017029302.1 3410 UTR 3 XP_016884791.1
XM_017029303.1 3410 Intron XP_016884792.1
XM_017029304.1 3410 UTR 3 XP_016884793.1
XM_017029305.1 3410 Intron XP_016884794.1
Gene
MPC1L
Gene Name
mitochondrial pyruvate carrier 1-like
There are no transcripts associated with this gene.

View Full Product Details