Product Details

SNP ID
rs141525139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:134819555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGAGAATGACATGATGAATTAG[A/G]ATGCTTTTGGCAAAAGCAGTAAAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM122C PubMed Links

Gene Details

Gene
FAM122C
Gene Name
family with sequence similarity 122C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170779.1 Intron NP_001164250.1
NM_001170780.1 Intron NP_001164251.1
NM_001170781.1 Intron NP_001164252.1
NM_001170782.1 Intron NP_001164253.1
NM_001170783.1 Intron NP_001164254.1
NM_001170784.1 Intron NP_001164255.1
NM_138819.3 Intron NP_620174.1
XM_005262382.1 Intron XP_005262439.1
XM_005262383.2 Intron XP_005262440.1
XM_005262384.4 Intron XP_005262441.1
XM_005262386.3 Intron XP_005262443.1
XM_005262387.3 Intron XP_005262444.1
XM_006724734.3 Intron XP_006724797.1
XM_006724735.1 Intron XP_006724798.1
XM_006724736.2 Intron XP_006724799.1
XM_011531295.1 Intron XP_011529597.1
XM_011531296.2 Intron XP_011529598.1
XM_011531297.2 Intron XP_011529599.1
XM_011531298.2 Intron XP_011529600.1
XM_011531299.1 Intron XP_011529601.1
XM_011531300.2 Intron XP_011529602.1
XM_011531301.2 Intron XP_011529603.1
XM_011531302.2 Intron XP_011529604.1
XM_011531305.2 Intron XP_011529607.1
XM_011531306.2 Intron XP_011529608.1
XM_017029308.1 Intron XP_016884797.1
XM_017029309.1 Intron XP_016884798.1
XM_017029310.1 Intron XP_016884799.1
XM_017029311.1 Intron XP_016884800.1

View Full Product Details