Product Details

SNP ID
rs142584263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:78656690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATAATGTCAAACTGGAGCTCTT[C/T]AACATGGTTTCCAATTTGAACTTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZCCHC5 PubMed Links

Gene Details

Gene
ZCCHC5
Gene Name
zinc finger CCHC-type containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152694.2 2027 UTR 3 NP_689907.1

View Full Product Details