Product Details
- SNP ID
-
rs142635989
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:74740143 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGTCCCAGACTCATCCTTGTGGA[C/T]CTGTTCTCGCTCCATGTGCTTTCCC
- Phenotype
-
MIM: 300524
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA2022
PubMed Links
Gene Details
- Gene
- KIAA2022
- Gene Name
- KIAA2022
View Full Product Details