Product Details

SNP ID
rs143008624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:38805491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAGATCGGCTTCGGCAATTGGG[A/G]CCACTGAGGCGTGGCGCCCGTGGCT
Phenotype
MIM: 300961
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MID1IP1 PubMed Links

Gene Details

Gene
MID1IP1
Gene Name
MID1 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098790.1 2744 Missense Mutation GAC,GGC D182G NP_001092260.1
NM_001098791.1 2744 Missense Mutation GAC,GGC D182G NP_001092261.1
NM_021242.5 2744 Missense Mutation GAC,GGC D182G NP_067065.1
Gene
MID1IP1-AS1
Gene Name
MID1IP1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details