Product Details

SNP ID
hCV175400144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120254068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAATAACACAGTGGCACAGGTCC[A/T]ATCTAACCCAGGCCCTGTTGCTATT
Phenotype
MIM: 300329
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NKAPP1 PubMed Links

Gene Details

Gene
NKAPP1
Gene Name
NFKB activating protein pseudogene 1
There are no transcripts associated with this gene.

Gene
TMEM255A
Gene Name
transmembrane protein 255A
There are no transcripts associated with this gene.

Gene
ZBTB33
Gene Name
zinc finger and BTB domain containing 33
There are no transcripts associated with this gene.

View Full Product Details