Product Details

SNP ID
rs143515418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119896266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGCAAAGAAGATCTGCTCACTC[C/T]GGTACTGGGGATTCTCCGGGGTGGG
Phenotype
MIM: 300462
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKAP14 PubMed Links

Gene Details

Gene
AKAP14
Gene Name
A-kinase anchoring protein 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008534.1 127 UTR 5 NP_001008534.1
NM_001008535.1 127 UTR 5 NP_001008535.1
NM_178813.5 127 UTR 5 NP_848928.1

View Full Product Details