Product Details

SNP ID
rs145529536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48600422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGGTGCCTGCCCCACCCACAGT[A/G]CTTCGCGTGCTACGGGGCCACACCC
Phenotype
MIM: 300512
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WDR13 PubMed Links

Gene Details

Gene
WDR13
Gene Name
WD repeat domain 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166426.1 785 Silent Mutation GTA,GTG V117V NP_001159898.1
NM_017883.4 785 Silent Mutation GTA,GTG V209V NP_060353.2
XM_006724539.1 785 Silent Mutation GTA,GTG V209V XP_006724602.1
XM_006724541.3 785 Silent Mutation GTA,GTG V117V XP_006724604.1
XM_011543947.2 785 Silent Mutation GTA,GTG V209V XP_011542249.1

View Full Product Details