Product Details

SNP ID
rs147523049
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48576521 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTGCTCTAGGTGGTGGGGACCA[A/G]GGCTATGGGAGTGGCAGGTATTATG
Phenotype
MIM: 300027
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RBM3 PubMed Links

Gene Details

Gene
RBM3
Gene Name
RNA binding motif (RNP1, RRM) protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006743.4 564 Silent Mutation CAA,CAG Q110Q NP_006734.1

View Full Product Details