Product Details

SNP ID
hCV175599184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49030447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGCATTCTGGGCAGGTCCCCCC[C/G]CTACATGGAACGTTGCTGCGCCTGG
Phenotype
MIM: 314310
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TFE3 PubMed Links

Gene Details

Gene
TFE3
Gene Name
transcription factor binding to IGHM enhancer 3
There are no transcripts associated with this gene.

View Full Product Details