Product Details

SNP ID
rs149841340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101080232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAAGCCCTTTTAGGGCTGAGGCA[A/G]GGAGGAACAGCCTCACACATTGCGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM35A PubMed Links

Gene Details

Gene
TMEM35A
Gene Name
transmembrane protein 35A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021637.2 Intron NP_067650.1

View Full Product Details