Product Details

SNP ID
rs149882611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48598837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCCCAGCGCTGCGTCGGCAGTA[C/T]CTGAGGCTTCGGGGGCAGCTGCTGG
Phenotype
MIM: 300512
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR13 PubMed Links

Gene Details

Gene
WDR13
Gene Name
WD repeat domain 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166426.1 320 UTR 5 NP_001159898.1
NM_017883.4 320 Silent Mutation TAC,TAT Y54Y NP_060353.2
XM_006724539.1 320 Silent Mutation TAC,TAT Y54Y XP_006724602.1
XM_006724541.3 320 UTR 5 XP_006724604.1
XM_011543947.2 320 Silent Mutation TAC,TAT Y54Y XP_011542249.1

View Full Product Details