Product Details

SNP ID
rs1140347
Assay Type
Functionally Tested
NCBI dbSNP Submissions
25
Location
Chr.6:32660245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATAGTCTCAGGAGTCAGTGCAG[G/A]AGCCCTGGAGAAGAGAGAAGAGCAT
Phenotype
MIM: 146880 MIM: 604305
Polymorphism
G/A, Transition Substitution
Allele Nomenclature
Literature Links
HLA-DQA1 PubMed Links

Gene Details

Gene
HLA-DQA1
Gene Name
major histocompatibility complex, class II, DQ alpha 1
There are no transcripts associated with this gene.

Gene
HLA-DQB1
Gene Name
major histocompatibility complex, class II, DQ beta 1
There are no transcripts associated with this gene.

Gene
HLA-DQB1-AS1
Gene Name
HLA-DQB1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details