Product Details

SNP ID
rs7624719
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:197755816 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGGTCATCTTTAAAAAATGGAG[C/G]CTTCTGTGATTATGGGAAGTAAGTA
Phenotype
MIM: 616933 MIM: 613516
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
FYTTD1 PubMed Links
Additional Information
For this assay, SNP(s) [rs148567326] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
FYTTD1
Gene Name
forty-two-three domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011537.2 245 Missense Mutation CCT,GCT P3A NP_001011537.2
NM_032288.6 245 Intron NP_115664.2
Gene
RUBCN
Gene Name
RUN and cysteine rich domain containing beclin 1 interacting protein
There are no transcripts associated with this gene.

View Full Product Details