Product Details

SNP ID
rs11171762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56161971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCCAGGCCATTATTAGCACTGC[A/C]TAAGCCTGTCCGTCACCTCAGCAGG
Phenotype
MIM: 609931 MIM: 609930 MIM: 601734
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC107987175 PubMed Links

Gene Details

Gene
LOC107987175
Gene Name
uncharacterized LOC107987175
There are no transcripts associated with this gene.

Gene
MYL6
Gene Name
myosin light chain 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021019.4 Intron NP_066299.2
NM_079423.3 Intron NP_524147.2
Gene
MYL6B
Gene Name
myosin light chain 6B
There are no transcripts associated with this gene.

Gene
SMARCC2
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005269101.1 Intron XP_005269158.1
XM_005269102.1 Intron XP_005269159.1
XM_005269103.1 Intron XP_005269160.1
XM_005269104.1 Intron XP_005269161.1
XM_011538693.2 Intron XP_011536995.1
XM_017019884.1 Intron XP_016875373.1
XM_017019885.1 Intron XP_016875374.1
XM_017019886.1 Intron XP_016875375.1
XM_017019887.1 Intron XP_016875376.1

View Full Product Details