Product Details

SNP ID
rs11695805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:47089737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTCTCTAGGTTTGGAGGAGTCC[G/T]GTATGCTCCCAGGATCCACTTTCTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C2orf61 PubMed Links

Gene Details

Gene
C2orf61
Gene Name
chromosome 2 open reading frame 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163561.1 Intron NP_001157033.1
NM_173649.2 Intron NP_775920.1

View Full Product Details