Product Details

SNP ID
rs11799219
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:9805308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAGGATTCCTCAGGGTGGCCTA[C/G]ATGCTGTAGGGGTGAGACTTACTGA
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
TTTY22 PubMed Links

Gene Details

Gene
TTTY22
Gene Name
testis-specific transcript, Y-linked 22 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details