Product Details

SNP ID
rs1204833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116251665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGAGGGGGAGGGAGAGGGGAGG[A/G]GGAGGGAGAGAAAGAGACAGAGGCA
Phenotype
MIM: 605942
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DSE PubMed Links

Gene Details

Gene
DSE
Gene Name
dermatan sulfate epimerase
There are no transcripts associated with this gene.

Gene
NT5DC1
Gene Name
5'-nucleotidase domain containing 1
There are no transcripts associated with this gene.

Gene
TSPYL4
Gene Name
TSPY like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021648.4 2434 UTR 3 NP_067680.3

View Full Product Details