Product Details

SNP ID
rs151341331
Assay Type
Functionally Tested
NCBI dbSNP Submissions
2
Location
Chr.6:31356186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTACCAGCGCGCTCCAGCTTGTC[T/C]TTCCCGTTCTCCAGGTATCTGCGGA
Phenotype
MIM: 142830
Polymorphism
T/C, Transition Substitution
Allele Nomenclature
Literature Links
HLA-B PubMed Links

Gene Details

Gene
HLA-B
Gene Name
major histocompatibility complex, class I, B
There are no transcripts associated with this gene.

Gene
MIR6891
Gene Name
microRNA 6891
There are no transcripts associated with this gene.

View Full Product Details