Product Details

SNP ID
rs181068440
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:1462652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGAGGAAGTGCTTAGAGATCGCT[C/G]GGCTGACAGAGGGCATGTCATGCCG
Phenotype
MIM: 612317
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATAD3B PubMed Links

Gene Details

Gene
ATAD3B
Gene Name
ATPase family, AAA domain containing 3B
There are no transcripts associated with this gene.

Gene
ATAD3C
Gene Name
ATPase family, AAA domain containing 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039211.2 2028 Missense Mutation CGG,GGG R345G NP_001034300.2

View Full Product Details