Product Details

SNP ID
rs181625518
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:6250547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTGGCCATCCTATCTAGGGCCA[A/C]AAAATCACCTCCATAGCCATAGTCC
Phenotype
MIM: 614269 MIM: 609971
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GPR153 PubMed Links

Gene Details

Gene
GPR153
Gene Name
G protein-coupled receptor 153
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207370.2 1403 Missense Mutation GTG,TTG V353L NP_997253.2
XM_011541434.2 1403 Missense Mutation GTG,TTG V353L XP_011539736.1
XM_017001250.1 1403 Missense Mutation GTG,TTG V353L XP_016856739.1
XM_017001251.1 1403 Missense Mutation GTG,TTG V353L XP_016856740.1
Gene
HES3
Gene Name
hes family bHLH transcription factor 3
There are no transcripts associated with this gene.

View Full Product Details