Product Details

SNP ID
rs181598526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42511850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAATAGAGTTTTGATTCATCATG[A/G]ATAATCTGTCATCAGAAGAAATTCA
Phenotype
MIM: 602534
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNAP23 PubMed Links

Gene Details

Gene
SNAP23
Gene Name
synaptosome associated protein 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003825.3 472 Missense Mutation AAT,GAT N2D NP_003816.2
NM_130798.2 472 Missense Mutation AAT,GAT N2D NP_570710.1
XM_006720725.2 472 Missense Mutation AAT,GAT N2D XP_006720788.1
XM_017022692.1 472 Missense Mutation AAT,GAT N2D XP_016878181.1
XM_017022693.1 472 Intron XP_016878182.1
XM_017022694.1 472 Intron XP_016878183.1

View Full Product Details