Product Details

SNP ID
rs181780563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:92969747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGATGTCAGAGAACGGCTCCTTC[G/T]CCAACTGCTGCGAGGCGGGCGGGTG
Phenotype
MIM: 600804
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MTNR1B PubMed Links

Gene Details

Gene
MTNR1B
Gene Name
melatonin receptor 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005959.3 125 Missense Mutation GCC,TCC A8S NP_005950.1
XM_011542839.2 125 Missense Mutation GCC,TCC A8S XP_011541141.1
XM_017017777.1 125 Intron XP_016873266.1

View Full Product Details