Product Details
- SNP ID
-
rs181290256
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:41184006 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGACAGGGAGGGAGTGTGGAAAC[C/T]ACAGTCTTTCTGATAGGATAAAAAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EXD1
PubMed Links
Gene Details
- Gene
- EXD1
- Gene Name
- exonuclease 3'-5' domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001286441.1 |
1463 |
Silent Mutation |
GTA,GTG |
V548V |
NP_001273370.1 |
NM_152596.3 |
1463 |
Silent Mutation |
GTA,GTG |
V490V |
NP_689809.2 |
XM_006720415.3 |
1463 |
Silent Mutation |
GTA,GTG |
V525V |
XP_006720478.1 |
XM_011521298.2 |
1463 |
Silent Mutation |
GTA,GTG |
V548V |
XP_011519600.1 |
XM_011521299.2 |
1463 |
Silent Mutation |
GTA,GTG |
V494V |
XP_011519601.1 |
XM_011521300.2 |
1463 |
Silent Mutation |
GTA,GTG |
V454V |
XP_011519602.1 |
XM_011521301.2 |
1463 |
Silent Mutation |
GTA,GTG |
V513V |
XP_011519603.1 |
XM_011521302.2 |
1463 |
Silent Mutation |
GTA,GTG |
V345V |
XP_011519604.1 |
XM_017021970.1 |
1463 |
Silent Mutation |
GTA,GTG |
V454V |
XP_016877459.1 |
XM_017021971.1 |
1463 |
Silent Mutation |
GTA,GTG |
V454V |
XP_016877460.1 |
View Full Product Details