Product Details

SNP ID
rs181906346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24204292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAAACAGGGTGAGACACAGCACG[A/T]CTTGGTAGAGATCCTTAATAAAGAA
Phenotype
MIM: 604464
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ITSN2 PubMed Links

Gene Details

Gene
ITSN2
Gene Name
intersectin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006277.2 5066 Missense Mutation GAC,GTC D1630V NP_006268.2
NM_019595.3 5066 Missense Mutation GAC,GTC D1603V NP_062541.3
NM_147152.2 5066 Intron NP_671494.2

View Full Product Details