Product Details

SNP ID
rs182336183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:444426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAAGTCTTGGGTGAAATGAGAA[A/C]ACATAGCTGAAAGAAACAAAAATAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ABCA11P PubMed Links

Gene Details

Gene
ABCA11P
Gene Name
ATP binding cassette subfamily A member 11, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF721
Gene Name
zinc finger protein 721
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133474.3 Intron NP_597731.2

View Full Product Details