Product Details
- SNP ID
-
rs182282798
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:35001370 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATATCTCCTCTATCTCTACTACC[A/G]CAGCTCAGGTTCAAACAGCCTCCTT
- Phenotype
-
MIM: 612471
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AGXT2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs5867261] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AGXT2
- Gene Name
- alanine--glyoxylate aminotransferase 2
View Full Product Details