Product Details

SNP ID
rs181994126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70657291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCTCCCTGTTGTTCAGAGGAAC[A/T]CTAAAAGAAAAGAAAGAAAACAAAG
Phenotype
MIM: 147790
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
JCHAIN PubMed Links

Gene Details

Gene
JCHAIN
Gene Name
joining chain of multimeric IgA and IgM
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144646.3 232 Silent Mutation ATA,ATT I63I NP_653247.1
XM_006714211.2 232 Silent Mutation ATA,ATT I63I XP_006714274.1
XM_006714212.2 232 Silent Mutation ATA,ATT I63I XP_006714275.1
XM_011531925.1 232 Silent Mutation ATA,ATT I63I XP_011530227.1
XM_011531926.1 232 Silent Mutation ATA,ATT I63I XP_011530228.1

View Full Product Details