Product Details

SNP ID
rs182024458
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:2185391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGAATAAAATGTTCCGATGATG[A/T]GGAAGCTCCTCTGATGCCTTTTTTC
Phenotype
MIM: 615183 MIM: 176982
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAAP20 PubMed Links

Gene Details

Gene
FAAP20
Gene Name
Fanconi anemia core complex associated protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146310.1 2230 Missense Mutation ACA,TCA T194S NP_001139782.1
NM_001256945.1 2230 Missense Mutation ACA,TCA T153S NP_001243874.1
NM_001256946.1 2230 Intron NP_001243875.1
NM_001256947.1 2230 Intron NP_001243876.1
NM_001282670.1 2230 Intron NP_001269599.1
NM_001282671.1 2230 Missense Mutation ACA,TCA T111S NP_001269600.1
NM_001282672.1 2230 Missense Mutation ACA,TCA T111S NP_001269601.1
NM_001282673.1 2230 UTR 3 NP_001269602.1
NM_182533.2 2230 Intron NP_872339.2
XM_006710419.3 2230 Intron XP_006710482.1
XM_006710421.3 2230 Intron XP_006710484.1
XM_011540914.2 2230 Intron XP_011539216.1
XM_011540921.2 2230 Intron XP_011539223.1
XM_011540922.1 2230 Intron XP_011539224.1
XM_017000553.1 2230 Intron XP_016856042.1
XM_017000554.1 2230 UTR 3 XP_016856043.1
XM_017000555.1 2230 Missense Mutation ACA,TCA T111S XP_016856044.1
XM_017000556.1 2230 Missense Mutation ACA,TCA T111S XP_016856045.1
XM_017000557.1 2230 Intron XP_016856046.1
Gene
LOC100506504
Gene Name
uncharacterized LOC100506504
There are no transcripts associated with this gene.

Gene
PRKCZ
Gene Name
protein kinase C zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033581.1 2230 UTR 3 NP_001028753.1
NM_001033582.1 2230 UTR 3 NP_001028754.1
NM_001242874.1 2230 UTR 3 NP_001229803.1
NM_002744.4 2230 UTR 3 NP_002735.3
XM_011541773.1 2230 Intron XP_011540075.1
XM_011541774.1 2230 Intron XP_011540076.1
XM_011541775.2 2230 Intron XP_011540077.1
XM_011541776.1 2230 Intron XP_011540078.1
XM_011541778.1 2230 Intron XP_011540080.1
XM_017001789.1 2230 Intron XP_016857278.1
XM_017001790.1 2230 UTR 3 XP_016857279.1
XM_017001791.1 2230 UTR 3 XP_016857280.1
XM_017001792.1 2230 Intron XP_016857281.1
XM_017001793.1 2230 Intron XP_016857282.1
XM_017001794.1 2230 Intron XP_016857283.1
XM_017001795.1 2230 Intron XP_016857284.1
XM_017001796.1 2230 UTR 3 XP_016857285.1
XM_017001797.1 2230 Intron XP_016857286.1
XM_017001798.1 2230 UTR 3 XP_016857287.1
XM_017001799.1 2230 UTR 3 XP_016857288.1
XM_017001800.1 2230 Intron XP_016857289.1
XM_017001801.1 2230 UTR 3 XP_016857290.1
XM_017001802.1 2230 Intron XP_016857291.1
XM_017001803.1 2230 Intron XP_016857292.1
XM_017001804.1 2230 UTR 3 XP_016857293.1
XM_017001805.1 2230 UTR 3 XP_016857294.1

View Full Product Details