Product Details

SNP ID
rs181841961
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:151806938 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCATTCTTTAACATCTTGTTTA[C/T]TAGCAAAAATCAGCAATCCAGCTTT
Phenotype
MIM: 608960
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL5A PubMed Links

Gene Details

Gene
ARL5A
Gene Name
ADP ribosylation factor like GTPase 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037174.1 437 Missense Mutation AAT,AGT N88S NP_001032251.1
NM_012097.3 437 Missense Mutation AAT,AGT N125S NP_036229.1
NM_177985.2 437 Missense Mutation AAT,AGT N88S NP_817114.2

View Full Product Details