Product Details

SNP ID
rs182470865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27497602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATTCTGACCACCATGGTACAGG[C/T]GGCTGGGAAAGTTCAGGAAGTGCTG
Phenotype
MIM: 611905 MIM: 600842 MIM: 607386
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FNDC4 PubMed Links

Gene Details

Gene
FNDC4
Gene Name
fibronectin type III domain containing 4
There are no transcripts associated with this gene.

Gene
GCKR
Gene Name
glucokinase (hexokinase 4) regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001486.3 323 Missense Mutation GCG,GTG A86V NP_001477.2
XM_011532763.1 323 Missense Mutation GCG,GTG A86V XP_011531065.1
XM_017003796.1 323 Intron XP_016859285.1
XM_017003797.1 323 Intron XP_016859286.1
Gene
IFT172
Gene Name
intraflagellar transport 172
There are no transcripts associated with this gene.

View Full Product Details